Assay Details
| Gene Symbol: Morc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010816.1 | NP_034946.1 | 5 - 6 | 418 | 94 | |
| GenBank mRNA | AF084945.1 | - | 5 - 6 | 418 | 94 | |
| BC030893.1 | - | 5 - 6 | 367 | 4487929 | 94 | |
More Information
Set Membership: |
|

Morc1
Genomic Map