Assay Details
| Gene Symbol: Per1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159367.1 | NP_001152839.1 | 10 - 11 | 1412 | 83 | |
| NM_011065.4 | NP_035195.2 | 10 - 11 | 1427 | 83 | ||
| GenBank mRNA | AB002108.1 | - | 9 - 10 | 1241 | 83 | |
| AF022992.1 | - | 10 - 11 | 1466 | 83 | ||
| AK081813.1 | - | 10 - 11 | 1425 | 83 | ||
| AK148202.1 | - | 10 - 11 | 1419 | 83 | ||
| AK154900.1 | - | 10 - 11 | 1408 | 83 | ||
| AK172958.1 | - | 10 - 11 | 1412 | 83 | ||
| BC039768.1 | - | 9 - 10 | 1312 | 4163009 | 83 | |
| BC091645.1 | - | 10 - 11 | 1414 | 30355259 | 83 | |
More Information
Set Membership: |
|

Per1
Genomic Map