Assay Details
| Gene Symbol: Nt5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011851.4 | NP_035981.1 | 7 - 8 | 1442 | 66 | |
| GenBank mRNA | AK028723.1 | - | 7 - 8 | 1412 | 66 | |
| AK029979.1 | - | 7 - 8 | 1412 | 66 | ||
| AK128997.1 | - | 7 - 8 | 1411 | 66 | ||
| AK154614.1 | - | 7 - 8 | 1419 | 66 | ||
| BC119268.1 | - | 7 - 8 | 1386 | 8734017 | 66 | |
| BC119270.1 | - | 7 - 8 | 1386 | 8734019 | 66 | |
| L12059.1 | - | 7 - 8 | 1365 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nt5e
Genomic Map