Assay Details
| Gene Symbol: Cib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011870.5 | NP_036000.1 | 3 - 4 | 437 | 72 | |
| GenBank mRNA | AB006463.1 | - | 3 - 4 | 218 | 72 | |
| AF173010.1 | - | 3 - 4 | 216 | 72 | ||
| AK010345.1 | - | 3 - 4 | 234 | 72 | ||
| AK027915.1 | - | 3 - 4 | 249 | 72 | ||
| AK152805.1 | - | 3 - 4 | 269 | 72 | ||
| AK165472.1 | - | 3 - 4 | 222 | 72 | ||
| BC003714.1 | - | 3 - 4 | 223 | 3599986 | 72 | |
| BC054385.1 | - | 3 - 4 | 242 | 6476137 | 72 | |
| CF104297.1 | - | 3 - 4 | 445 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
positive regulation of cell-matrix adhesion
response to ischemia
negative regulation of protein kinase activity
apoptotic process
cellular response to DNA damage stimulus
negative regulation of microtubule depolymerization
cell cycle
endomitotic cell cycle
cell adhesion
spermatogenesis
spermatid development
positive regulation of cell proliferation
negative regulation of cell proliferation
negative regulation of neuron projection development
cell differentiation
platelet formation
positive regulation of cell growth
positive regulation of cell migration
cytoplasmic microtubule organization
positive regulation of cell adhesion mediated by integrin
thrombopoietin-mediated signaling pathway
regulation of cell proliferation
negative regulation of apoptotic process
positive regulation of catalytic activity
negative regulation of megakaryocyte differentiation
positive regulation of NF-kappaB transcription factor activity
cell division
regulation of cell division
negative regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
positive regulation of calcineurin-NFAT signaling cascade
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
negative regulation of protein serine/threonine kinase activity
positive regulation of protein serine/threonine kinase activity
positive regulation of establishment of protein localization to plasma membrane
positive regulation of cell migration involved in sprouting angiogenesis
positive regulation of protein targeting to membrane
positive regulation of substrate adhesion-dependent cell spreading
cellular response to nerve growth factor stimulus
positive regulation of male germ cell proliferation
calcium ion binding
protein binding
calcium-dependent protein kinase inhibitor activity
Ras GTPase binding
protein serine/threonine kinase inhibitor activity
protein anchor
ion channel binding
metal ion binding
angiogenesis
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
positive regulation of cell-matrix adhesion
response to ischemia
negative regulation of protein kinase activity
apoptotic process
cellular response to DNA damage stimulus
negative regulation of microtubule depolymerization
cell cycle
endomitotic cell cycle
cell adhesion
spermatogenesis
spermatid development
positive regulation of cell proliferation
negative regulation of cell proliferation
negative regulation of neuron projection development
cell differentiation
platelet formation
positive regulation of cell growth
positive regulation of cell migration
cytoplasmic microtubule organization
positive regulation of cell adhesion mediated by integrin
thrombopoietin-mediated signaling pathway
regulation of cell proliferation
negative regulation of apoptotic process
positive regulation of catalytic activity
negative regulation of megakaryocyte differentiation
positive regulation of NF-kappaB transcription factor activity
cell division
regulation of cell division
negative regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
positive regulation of calcineurin-NFAT signaling cascade
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
negative regulation of protein serine/threonine kinase activity
positive regulation of protein serine/threonine kinase activity
positive regulation of establishment of protein localization to plasma membrane
positive regulation of cell migration involved in sprouting angiogenesis
positive regulation of protein targeting to membrane
positive regulation of substrate adhesion-dependent cell spreading
cellular response to nerve growth factor stimulus
positive regulation of male germ cell proliferation
calcium ion binding
protein binding
calcium-dependent protein kinase inhibitor activity
Ras GTPase binding
protein serine/threonine kinase inhibitor activity
protein anchor
ion channel binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25265481 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25265477 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25265480 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25265479 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982683 | BC003714.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982683 | BC054385.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982683 | AK152805.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982683 | NM_011870.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1982683 | AK010345.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969893 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835124 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434387 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155724 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982683 | BC003714.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982683 | BC054385.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982683 | AK152805.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982683 | NM_011870.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1982683 | AK010345.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123384 | AK165472.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123384 | BC003714.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123384 | BC054385.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123384 | AK152805.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P123384 | NM_011870.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175454 | AK165472.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175454 | BC003714.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175454 | BC054385.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175454 | AK152805.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P175454 | NM_011870.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265477 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25265481 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155724 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06255807 | - | - |
| Whole Mouse Genome G4122A | A_52_P123384 | AK165472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123384 | BC003714.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123384 | BC054385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123384 | AK152805.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P123384 | NM_011870.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175454 | AK165472.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175454 | BC003714.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175454 | BC054385.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175454 | AK152805.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P175454 | NM_011870.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100876 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106225 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155724 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06237985 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06237995 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06237999 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06237981 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25265477 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25265480 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25265479 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25265481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25265475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06237973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06417267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06438695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265477 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265448 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06361099 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06101192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265456 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25265461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10564871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10564874 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425856_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448564_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10564869 | - | - |
| Mouse Genome 430 2.0 Array | 1448564_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425856_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425856_at | - | - |
| Mouse Genome 430 2.0 Array | 1448564_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 23991_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 23991_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700030299.mm.1 | NM_011870.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700030299.mm.1 | TC0700003586.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425856_at | - | - |
| Mouse Expression Set 430 | 1448564_PM_at | - | - |
| Mouse Expression Set 430 | 1425856_PM_at | - | - |
| Mouse Expression Set 430 | 1448564_at | - | - |
| Murine Genome U74v2 Set | 96957_at | - | - |
| Murine Genome U74v2 Set | 96958_PM_at | - | - |
| Murine Genome U74v2 Set | 96957_PM_at | - | - |
| Murine Genome U74v2 Set | 96958_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425856_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425856_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448564_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448564_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448564_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425856_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448564_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425856_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 775784 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 7100722 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007100722 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2621844 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2621844 | - | - |
| mouse-6 v1.1 (Illumina) | scl31132.7.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7100722 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31132.7.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7100722 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2621847 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2621847 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31132.7.1_7-S | - | - |

Cib1
Genomic Map