Assay Details
| Gene Symbol: Cib1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291275.1 | NP_001278204.1 | 5 - 6 | 696 | 77 | |
| NM_001291276.1 | NP_001278205.1 | 3 - 4 | 567 | 77 | ||
| NM_011870.5 | NP_036000.1 | 5 - 6 | 711 | 77 | ||
| GenBank mRNA | AB006463.1 | - | 5 - 6 | 492 | 77 | |
| AF173010.1 | - | 5 - 6 | 490 | 77 | ||
| AK010345.1 | - | 5 - 6 | 508 | 77 | ||
| AK027915.1 | - | 5 - 6 | 523 | 77 | ||
| AK088450.1 | - | 3 - 4 | 382 | 77 | ||
| AK152805.1 | - | 5 - 6 | 543 | 77 | ||
| AK165472.1 | - | 5 - 6 | 496 | 77 | ||
| BC003714.1 | - | 5 - 6 | 497 | 3599986 | 77 | |
| BC054385.1 | - | 5 - 6 | 516 | 6476137 | 77 | |
More Information
Set Membership: |
|

Cib1
Genomic Map