Assay Details
| Gene Symbol: Tuba8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017379.1 | NP_059075.1 | 1 - 2 | 72 | 62 | |
| GenBank mRNA | AJ245923.1 | - | 1 - 2 | 72 | 62 | |
| AK032157.1 | - | 45 | 62 | |||
| BC017631.1 | - | 1 - 2 | 51 | 4507364 | 62 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ245923.1,BC017631.1,NM_017379.1 transcripts.
Set Membership: |
|

Tuba8
Genomic Map