Assay Details
| Gene Symbol: Slc7a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017394.4 | NP_059090.3 | 4 - 5 | 747 | 67 | |
| GenBank mRNA | AB026688.1 | - | 4 - 5 | 661 | 67 | |
| AK005282.1 | - | 2 - 3 | 144 | 67 | ||
| AK031867.1 | - | 4 - 5 | 715 | 67 | ||
| BC054765.1 | - | 4 - 5 | 690 | 6414759 | 67 | |
More Information
Set Membership: |
|

Slc7a10
Genomic Map