Assay Details
| Gene Symbol: Slc7a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017394.4 | NP_059090.3 | 5 - 6 | 907 | 65 | |
| GenBank mRNA | AB026688.1 | - | 5 - 6 | 821 | 65 | |
| AK005282.1 | - | 3 - 4 | 304 | 65 | ||
| AK031867.1 | - | 5 - 6 | 875 | 65 | ||
| BC054765.1 | - | 5 - 6 | 850 | 6414759 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a10
Genomic Map