Assay Details
| Gene Symbol: Abhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018811.6 | NP_061281.3 | 4 - 5 | 545 | 85 | |
| GenBank mRNA | AF201730.1 | - | 4 - 5 | 534 | 85 | |
| AF546701.1 | - | 4 - 5 | 546 | 85 | ||
| AK008690.1 | - | 4 - 5 | 555 | 85 | ||
| AK016296.1 | - | 2 - 3 | 206 | 85 | ||
| AK053460.1 | - | 4 - 5 | 654 | 85 | ||
| AK139526.1 | - | 4 - 5 | 554 | 85 | ||
| AK153642.1 | - | 4 - 5 | 592 | 85 | ||
| BC027098.1 | - | 4 - 5 | 515 | 5356667 | 85 | |
| BC089477.1 | - | 4 - 5 | 525 | 6741521 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abhd2
Genomic Map