Assay Details
| Gene Symbol: Abhd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018811.6 | NP_061281.3 | 9 - 10 | 1172 | 72 | |
| GenBank mRNA | AF201730.1 | - | 9 - 10 | 1161 | 72 | |
| AF546701.1 | - | 9 - 10 | 1173 | 72 | ||
| AK008690.1 | - | 9 - 10 | 1182 | 72 | ||
| AK016296.1 | - | 7 - 8 | 833 | 72 | ||
| AK139526.1 | - | 9 - 10 | 1181 | 72 | ||
| AK153642.1 | - | 9 - 10 | 1219 | 72 | ||
| BC027098.1 | - | 9 - 10 | 1142 | 5356667 | 72 | |
| BC089477.1 | - | 9 - 10 | 1152 | 6741521 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abhd2
Genomic Map