Assay Details
| Gene Symbol: Rabep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291141.1 | NP_001278070.1 | 10 - 11 | 1869 | 124 | |
| NM_001291142.1 | NP_001278071.1 | 10 - 11 | 1878 | 124 | ||
| NM_001291143.1 | NP_001278072.1 | 12 - 13 | 2069 | 124 | ||
| NM_019400.3 | NP_062273.2 | 11 - 12 | 1998 | 124 | ||
| GenBank mRNA | AB001750.1 | - | 12 - 13 | 1905 | 124 | |
| AF248489.1 | - | 10 - 11 | 1664 | 124 | ||
| AF248490.1 | - | 10 - 11 | 1655 | 124 | ||
| AK134587.1 | - | 11 - 12 | 1999 | 124 | ||
| BC003921.1 | - | 2 - 3 | 619 | 124 | ||
| BC060166.1 | - | 11 - 12 | 2016 | 124 | ||
| BC080274.1 | - | 11 - 12 | 1970 | 124 | ||
| BC129853.1 | - | 11 - 12 | 1817 | 40109688 | 124 | |
| BC129854.1 | - | 11 - 12 | 1817 | 40109689 | 124 | |
| D86066.1 | - | 11 - 12 | 1834 | 124 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabep1
Genomic Map