Assay Details
| Gene Symbol: Sae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285891.1 | NP_001272820.1 | 5 - 6 | 779 | 80 | |
| NM_001285892.1 | NP_001272821.1 | 5 - 6 | 779 | 80 | ||
| NM_019748.3 | NP_062722.1 | 5 - 6 | 779 | 80 | ||
| GenBank mRNA | AB024303.1 | - | 5 - 6 | 667 | 80 | |
| AK010313.1 | - | 5 - 6 | 656 | 80 | ||
| AK011772.1 | - | 5 - 6 | 690 | 80 | ||
| AK011783.1 | - | 2 - 3 | 126 | 80 | ||
| AK087556.1 | - | 5 - 6 | 670 | 80 | ||
| AK090012.1 | - | 5 - 6 | 666 | 80 | ||
| AK154139.1 | - | 5 - 6 | 692 | 80 | ||
| AK159672.1 | - | 5 - 6 | 681 | 80 | ||
| AK162789.1 | - | 5 - 6 | 667 | 80 | ||
| BC068164.1 | - | 5 - 6 | 668 | 30359189 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sae1
Genomic Map