Assay Details
| Gene Symbol: Akap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB028920.1 | - | 3 - 4 | 126 | 76 | |
| AK004801.1 | - | 3 - 4 | 132 | 76 | ||
| AK089092.1 | - | 3 - 4 | 139 | 76 | ||
| BC042602.1 | - | 3 - 4 | 122 | 76 | ||
| BC060170.1 | - | 3 - 4 | 117 | 76 | ||
| BC060209.1 | - | 3 - 4 | 223 | 76 | ||
| BC078448.1 | - | 3 - 4 | 110 | 76 | ||
| BC125521.1 | - | 3 - 4 | 122 | 40130036 | 76 | |
| BC137746.1 | - | 3 - 4 | 122 | 8860765 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap8
Genomic Map