Assay Details
| Gene Symbol: Akap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB028920.1 | - | 10 - 11 | 1331 | 80 | |
| AK004801.1 | - | 10 - 11 | 1337 | 80 | ||
| AK089092.1 | - | 10 - 11 | 1344 | 80 | ||
| BC042602.1 | - | 10 - 11 | 2214 | 80 | ||
| BC054433.1 | - | 8 - 9 | 1911 | 80 | ||
| BC060170.1 | - | 10 - 11 | 2211 | 80 | ||
| BC060209.1 | - | 9 - 10 | 1827 | 80 | ||
| BC078448.1 | - | 9 - 10 | 1717 | 80 | ||
| BC125521.1 | - | 10 - 11 | 1327 | 40130036 | 80 | |
| BC137746.1 | - | 10 - 11 | 1327 | 8860765 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akap8
Genomic Map