Assay Details
| Gene Symbol: Piwil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021308.1 | NP_067283.1 | 13 - 14 | 1736 | 101 | |
| GenBank mRNA | AB032605.1 | - | 13 - 14 | 1736 | 101 | |
| AF285586.1 | - | 4 - 5 | 465 | 101 | ||
| AK030116.1 | - | 13 - 14 | 1680 | 101 | ||
| BC138444.1 | - | 13 - 14 | 1724 | 8861464 | 101 | |
| BC145717.1 | - | 13 - 14 | 1724 | 40131020 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Piwil2
Genomic Map