Assay Details
| Gene Symbol: Ppp2r3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021529.3 | NP_067504.2 | 1 - 2 | 86 | 76 | |
| GenBank mRNA | AB041577.1 | - | 1 - 2 | 75 | 76 | |
| AK028063.1 | - | 1 - 2 | 51 | 76 | ||
| AK033405.1 | - | 1 - 2 | 84 | 76 | ||
| AK163192.1 | - | 85 | 76 | |||
| AK168298.1 | - | 1 - 2 | 87 | 76 | ||
| BC024754.1 | - | 1 - 2 | 49 | 5375405 | 76 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB041577.1,BC024754.1,NM_021529.3 transcripts.
Set Membership: |
|

Ppp2r3c
Genomic Map