Assay Details
| Gene Symbol: Actn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021895.2 | NP_068695.1 | 2 - 3 | 376 | 61 | |
| GenBank mRNA | AJ289242.2 | - | 2 - 3 | 376 | 61 | |
| AK145322.1 | - | 2 - 3 | 429 | 61 | ||
| AK150044.1 | - | 2 - 3 | 378 | 61 | ||
| BC013616.1 | - | 2 - 3 | 385 | 3157024 | 61 | |
| BC027032.1 | - | 2 - 3 | 327 | 61 | ||
| BC087554.1 | - | 2 - 3 | 329 | 5351470 | 61 | |
| DQ303411.1 | - | 2 - 3 | 279 | 61 | ||
| U41415.1 | - | 1 - 2 | 63 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actn4
Genomic Map