Assay Details
| Gene Symbol: Actn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021895.2 | NP_068695.1 | 15 - 16 | 1978 | 65 | |
| GenBank mRNA | AJ289242.2 | - | 15 - 16 | 1978 | 65 | |
| AK030237.1 | - | 8 - 9 | 1148 | 65 | ||
| AK145322.1 | - | 15 - 16 | 2031 | 65 | ||
| AK150044.1 | - | 15 - 16 | 1980 | 65 | ||
| BC013616.1 | - | 15 - 16 | 1987 | 3157024 | 65 | |
| BC027032.1 | - | 15 - 16 | 1927 | 65 | ||
| BC087554.1 | - | 15 - 16 | 1931 | 5351470 | 65 | |
| DQ303411.1 | - | 15 - 16 | 1881 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Actn4
Genomic Map