Assay Details
| Gene Symbol: Actn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021895.2 | NP_068695.1 | 16 - 17 | 2109 | 85 | |
| GenBank mRNA | AJ289242.2 | - | 16 - 17 | 2109 | 85 | |
| AK030237.1 | - | 9 - 10 | 1279 | 85 | ||
| AK145322.1 | - | 16 - 17 | 2162 | 85 | ||
| AK150044.1 | - | 16 - 17 | 2111 | 85 | ||
| BC013616.1 | - | 16 - 17 | 2118 | 3157024 | 85 | |
| BC027032.1 | - | 16 - 17 | 2058 | 85 | ||
| BC087554.1 | - | 16 - 17 | 2062 | 5351470 | 85 | |
| DQ303411.1 | - | 16 - 17 | 2012 | 85 | ||
More Information
Set Membership: |
|

Actn4
Genomic Map