Assay Details
| Gene Symbol: Gtf2h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022011.4 | NP_071294.3 | 11 - 12 | 945 | 69 | |
| GenBank mRNA | AB221674.1 | - | 10 - 11 | 760 | 69 | |
| AK154739.1 | - | 10 - 11 | 1267 | 69 | ||
| BC016231.1 | - | 11 - 12 | 900 | 4488798 | 69 | |
| BC053382.1 | - | 11 - 12 | 909 | 30047823 | 69 | |
| DV662169.1 | - | 9 - 10 | 685 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf2h2
Genomic Map