Assay Details
| Gene Symbol: Clstn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022319.2 | NP_071714.2 | 7 - 8 | 1420 | 78 | |
| GenBank mRNA | AJ278069.1 | - | 7 - 8 | 1368 | 78 | |
| AK220500.1 | - | 7 - 8 | 1434 | 78 | ||
| BC063058.1 | - | 7 - 8 | 1420 | 5705591 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clstn2
Genomic Map