Assay Details
| Gene Symbol: Clstn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022319.2 | NP_071714.2 | 13 - 14 | 2419 | 78 | |
| GenBank mRNA | AJ278069.1 | - | 13 - 14 | 2367 | 78 | |
| AK048369.1 | - | 6 - 7 | 953 | 78 | ||
| AK220500.1 | - | 13 - 14 | 2433 | 78 | ||
| BC063058.1 | - | 13 - 14 | 2419 | 5705591 | 78 | |
More Information
Set Membership: |
|

Clstn2
Genomic Map