Assay Details
| Gene Symbol: Clstn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290989.1 | NP_001277918.1 | 10 - 11 | 1762 | 74 | |
| NM_023051.5 | NP_075538.1 | 11 - 12 | 1792 | 74 | ||
| GenBank mRNA | AJ289016.1 | - | 11 - 12 | 1623 | 74 | |
| AK034544.1 | - | 3 - 4 | 335 | 74 | ||
| AK173059.1 | - | 10 - 11 | 1802 | 74 | ||
| AK203469.1 | - | 2 - 3 | 113 | 74 | ||
| BC029027.1 | - | 6 - 7 | 877 | 74 | ||
| BC053843.1 | - | 10 - 11 | 1731 | 6315355 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clstn1
Genomic Map