Assay Details
| Gene Symbol: Clstn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290989.1 | NP_001277918.1 | 12 - 13 | 2065 | 56 | |
| NM_023051.5 | NP_075538.1 | 13 - 14 | 2095 | 56 | ||
| GenBank mRNA | AJ289016.1 | - | 13 - 14 | 1926 | 56 | |
| AK034544.1 | - | 5 - 6 | 638 | 56 | ||
| AK173059.1 | - | 12 - 13 | 2105 | 56 | ||
| BC029027.1 | - | 8 - 9 | 1180 | 56 | ||
| BC053843.1 | - | 12 - 13 | 2034 | 6315355 | 56 | |
| CX566257.1 | - | 2 - 3 | 165 | 56 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Clstn1
Genomic Map