Assay Details
| Gene Symbol: Clstn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290989.1 | NP_001277918.1 | 13 - 14 | 2296 | 75 | |
| NM_023051.5 | NP_075538.1 | 14 - 15 | 2326 | 75 | ||
| GenBank mRNA | AJ289016.1 | - | 14 - 15 | 2157 | 75 | |
| AK034544.1 | - | 6 - 7 | 868 | 75 | ||
| AK173059.1 | - | 13 - 14 | 2336 | 75 | ||
| BC029027.1 | - | 9 - 10 | 1411 | 75 | ||
| BC053843.1 | - | 13 - 14 | 2265 | 6315355 | 75 | |
| CX566257.1 | - | 3 - 4 | 396 | 75 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Clstn1
Genomic Map