Assay Details
Gene Symbol: Atp6v0e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025272.2 | NP_079548.1 | 2 - 3 | 234 | 66 | |
GenBank mRNA | AK007610.1 | - | 2 - 3 | 213 | 66 | |
AK008464.1 | - | 2 - 3 | 214 | 66 | ||
AK013229.1 | - | 2 - 3 | 195 | 66 | ||
AK017760.1 | - | 2 - 3 | 236 | 66 | ||
AK030335.1 | - | 2 - 3 | 186 | 66 | ||
AK134836.1 | - | 2 - 3 | 218 | 66 | ||
AK159669.1 | - | 2 - 3 | 222 | 66 | ||
AK168015.1 | - | 2 - 3 | 203 | 66 | ||
BC024733.1 | - | 2 - 3 | 198 | 5374017 | 66 | |
BY707658.1 | - | 2 - 3 | 210 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
