Assay Details
| Gene Symbol: Cystm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002512.1 | - | 293 | 97 | ||
| AK003470.1 | - | 121 | 97 | |||
| AK028217.1 | - | 291 | 97 | |||
| AK167679.1 | - | 325 | 97 | |||
| BC028765.1 | - | 347 | 1195860 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cystm1
Genomic Map