Assay Details
| Gene Symbol: Sdhaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025333.4 | NP_079609.2 | 3 - 4 | 482 | 92 | |
| GenBank mRNA | AK013454.1 | - | 3 - 4 | 387 | 92 | |
| AK075594.1 | - | 3 - 4 | 387 | 92 | ||
| AK160175.1 | - | 1 - 2 | 115 | 92 | ||
| AK160957.1 | - | 3 - 4 | 389 | 92 | ||
| AK167228.1 | - | 3 - 4 | 418 | 92 | ||
| BC029630.1 | - | 2 - 3 | 201 | 92 | ||
| BC038924.1 | - | 2 - 3 | 450 | 92 | ||
| BC048913.1 | - | 3 - 4 | 215 | 92 | ||
| BC085277.1 | - | 3 - 4 | 384 | 30441476 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951137 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074062 | BC038924.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074062 | BC048913.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074062 | BC085277.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074062 | AK160957.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2074062 | AK167228.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 938325 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22387 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385243 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 324091 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441133 | AF079853.2 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441133 | AK080957.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441133 | AK152776.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441133 | BC055376.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441133 | NM_001085518.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441133 | NM_172980.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441133 | XM_001480396.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193784 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831670 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956862 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170879 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90856 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147545 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174354 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074062 | BC038924.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074062 | BC048913.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074062 | BC085277.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074062 | AK160957.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2074062 | AK167228.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31951136 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09104016 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147545 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09104037 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174354 | - | - |
| Whole Mouse Genome G4122A | A_52_P367681 | BC038924.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367681 | BC048913.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367681 | BC085277.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367681 | AK160957.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367681 | AK167228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P484691 | BC085277.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P484691 | BC029630.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P484691 | AK160175.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P484691 | NM_025333.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P484691 | AK075594.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P209973 | AK083578.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P367688 | AK013454.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128192 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174354 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147545 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14375155 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14375156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951135 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31951137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31950999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31951047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14260566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14468499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14311672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951135 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951072 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14468493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14510278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951069 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951062 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14231439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14325182 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14432723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951078 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951052 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14375155 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31951048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14468492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14468497 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465988 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465989 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465986 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465987 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452941_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428880_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465980 | - | - |
| Mouse Genome 430 2.0 Array | 1452941_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1428880_at | - | - |
| Mouse Genome 430 2.0 Array | 1428880_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452941_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66072_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66072_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900010330.mm.1 | NM_025333.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010330.mm.1 | NM_025333.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010330.mm.1 | TC1900001104.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900010331.mm.1 | NM_025333.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010331.mm.1 | NM_025333.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010331.mm.1 | TC1900001104.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900010335.mm.1 | NM_025333.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010335.mm.1 | NM_025333.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900010335.mm.1 | TC1900001104.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428880_at | - | - |
| Mouse Expression Set 430 | 1428880_PM_at | - | - |
| Mouse Expression Set 430 | 1452941_at | - | - |
| Mouse Expression Set 430 | 1452941_PM_at | - | - |
| Murine Genome U74v2 Set | 103097_PM_at | - | - |
| Murine Genome U74v2 Set | 103097_at | - | - |
| Murine Genome U74v2 Set | 130498_f_at | - | - |
| Murine Genome U74v2 Set | 166987_PM_r_at | - | - |
| Murine Genome U74v2 Set | 166987_r_at | - | - |
| Murine Genome U74v2 Set | 130498_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428880_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452941_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452941_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428880_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428880_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452941_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428880_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452941_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 624649 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2940673 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940673 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2596016 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2596016 | - | - |
| mouse-6 v1.1 (Illumina) | scl52754.4_241-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl52754.4_241-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2596016 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2596016 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl52754.4_241-S | - | - |

Sdhaf2
Genomic Map