Assay Details
| Gene Symbol: Prr13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170911.1 | NP_001164382.1 | 2 - 3 | 131 | 79 | |
| NM_025385.3 | NP_079661.1 | 3 - 4 | 249 | 79 | ||
| GenBank mRNA | AK002430.1 | - | 2 - 3 | 99 | 79 | |
| AK003850.1 | - | 3 - 4 | 246 | 79 | ||
| AK008611.1 | - | 1 - 2 | 71 | 79 | ||
| AK008894.1 | - | 3 - 4 | 209 | 79 | ||
| AK154583.1 | - | 3 - 4 | 206 | 79 | ||
| BC016234.1 | - | 2 - 3 | 395 | 4503179 | 79 | |
| BY285374.1 | - | 3 - 4 | 251 | 79 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK002430.1,AK003850.1,AK008611.1,AK008894.1,AK154583.1,BC016234.1,NM_001170911.1,NM_025385.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069217 | AK008894.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069217 | AK154583.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069217 | AK181565.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069217 | BC016234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2069217 | NM_001170911.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233949 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123023 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069217 | AK008894.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069217 | AK154583.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069217 | AK181565.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069217 | BC016234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2069217 | NM_001170911.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497945 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30497954 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123023 | - | - |
| Whole Mouse Genome G4122A | A_51_P355472 | AK008894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P355472 | AK154583.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P355472 | BC016234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P355472 | NM_001170911.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P355472 | NM_025385.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100874 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123023 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11856134 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11856129 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12078204 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12024302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30498002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30497950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497945 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497943 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12325919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497962 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12156948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12142242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11980293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30497972 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10427237 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427240 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427236 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427238 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10427239 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1423686_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10427235 | - | - |
| Mouse Genome 430 2.0 Array | 1423686_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1423686_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66151_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66151_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500008858.mm.1 | NM_001170911.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008858.mm.1 | NM_001170911.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008858.mm.1 | NM_025385.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008858.mm.1 | NM_025385.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008858.mm.1 | TC1500001101.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008860.mm.1 | NM_001170911.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008860.mm.1 | NM_001170911.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008860.mm.1 | NM_025385.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008860.mm.1 | NM_025385.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008860.mm.1 | TC1500001101.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500008859.mm.1 | NM_001170911.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008859.mm.1 | NM_001170911.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008859.mm.1 | NM_025385.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008859.mm.1 | NM_025385.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500008859.mm.1 | TC1500001101.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1423686_a_at | - | - |
| Mouse Expression Set 430 | 1423686_PM_a_at | - | - |
| Murine Genome U74v2 Set | 96278_PM_at | - | - |
| Murine Genome U74v2 Set | 96278_at | - | - |
| Murine Genome U74v2 Set | 169061_r_at | - | - |
| Murine Genome U74v2 Set | 169061_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423686_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1423686_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423686_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1423686_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2850452 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850452 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2657516 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2657516 | - | - |
| mouse-6 v1.1 (Illumina) | scl066151.5_253-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850452 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl066151.5_253-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850452 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2657516 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2657516 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl066151.5_253-S | - | - |

Prr13
Genomic Map