Assay Details
| Gene Symbol: Acer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025408.2 | NP_079684.2 | 2 - 3 | 293 | 67 | |
| GenBank mRNA | AK004287.1 | - | 2 - 3 | 294 | 67 | |
| AK011668.1 | - | 2 - 3 | 274 | 67 | ||
| AK017361.1 | - | 198 | 67 | |||
| AK080977.1 | - | 2 - 3 | 271 | 67 | ||
| AK150629.1 | - | 2 - 3 | 254 | 67 | ||
| AK162403.1 | - | 2 - 3 | 309 | 67 | ||
| BC023924.1 | - | 2 - 3 | 297 | 5324078 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acer3
Genomic Map