Assay Details
| Gene Symbol: Them5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025416.3 | NP_079692.1 | 5 - 6 | 864 | 73 | |
| GenBank mRNA | AK004155.1 | - | 5 - 6 | 865 | 73 | |
| AK009228.1 | - | 5 - 6 | 863 | 73 | ||
| BC046437.1 | - | 5 - 6 | 846 | 3969280 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | AK004155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | AK009228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | BC046437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | NM_025416.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | AK003517.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | AK004155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | AK009228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | BC046437.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | NM_025416.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267441 | AK003517.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682409 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109285 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | AK004155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | AK009228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | BC046437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | NM_025416.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | AK003517.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | AK004155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | AK009228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | BC046437.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | NM_025416.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267441 | AK003517.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338441 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22338434 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04992284 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109285 | - | - |
| Whole Mouse Genome G4122A | A_51_P267441 | AK004155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267441 | AK009228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267441 | BC046437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267441 | NM_025416.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267441 | AK003517.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267441 | AK004155.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267441 | AK009228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267441 | BC046437.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267441 | NM_025416.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267441 | AK003517.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104999 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139849 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109285 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02584046 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02584061 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02584047 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02584066 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02584058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22338437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338387 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02302697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03045984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02584063 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03185048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02912045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02738316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02584061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02919414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02584064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02584057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03185043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338433 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02584046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02738321 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02562769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02302690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02456967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338459 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02584041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338383 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338405 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02302694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02912046 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02653353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02919424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02302692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338408 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338407 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03185051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338382 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338447 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338424 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22338400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02738328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02584050 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10494018 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494021 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494019 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494020 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494022 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494017 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429354_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453801_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431211_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494016 | - | - |
| Mouse Genome 430 2.0 Array | 1429354_at | - | - |
| Mouse Genome 430 2.0 Array | 1431211_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1453801_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431211_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1429354_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453801_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66198_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66198_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300006582.mm.1 | NM_025416.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006582.mm.1 | NM_025416.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300006582.mm.1 | TC0300000862.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431211_PM_s_at | - | - |
| Mouse Expression Set 430 | 1453801_PM_at | - | - |
| Mouse Expression Set 430 | 1453801_at | - | - |
| Mouse Expression Set 430 | 1431211_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429354_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453801_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431211_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429354_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431211_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453801_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 916266 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 7000170 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7000170 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007000170 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2712007 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2712007 | - | - |
| mouse-6 v1.1 (Illumina) | scl22908.1.21_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7000170 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22908.1.21_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7000170 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2712007 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2946970 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2712007 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2946970 | - | - |

Them5
Genomic Map