Assay Details
| Gene Symbol: Med7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025426.3 | NP_079702.3 | 1 - 2 | 234 | 113 | |
| GenBank mRNA | AK004354.1 | - | 1 - 2 | 199 | 112 | |
| AK012799.1 | - | 1 - 2 | 214 | 113 | ||
| AK147739.1 | - | 1 - 2 | 224 | 113 | ||
| AK167407.1 | - | 1 - 2 | 194 | 113 | ||
| BC011333.1 | - | 1 - 2 | 221 | 3993084 | 113 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK004354.1,AK012799.1,AK147739.1,AK167407.1,BC011333.1,NM_025426.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767874 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767867 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767870 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767873 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767868 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960281 | AK009549.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960281 | CK788351.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960281 | BC011333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960281 | AK167407.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1960281 | AK010357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | BC011333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | AK167407.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | AK010357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | AK012799.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | AK144918.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | BC011333.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | AK167407.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | AK010357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | AK012799.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P327467 | AK144918.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50195 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 565448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 21811 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125812 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139478 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | BC011333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | AK167407.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | AK010357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | AK012799.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | AK144918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | BC011333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | AK167407.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | AK010357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | AK012799.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P327467 | AK144918.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960281 | AK009549.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960281 | CK788351.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960281 | BC011333.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960281 | AK167407.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1960281 | AK010357.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767872 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767862 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767868 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27767842 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125812 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139478 | - | - |
| Whole Mouse Genome G4122A | A_52_P327467 | BC011333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327467 | AK167407.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327467 | AK010357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327467 | AK012799.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327467 | AK144918.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327467 | BC011333.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327467 | AK167407.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327467 | AK010357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327467 | AK012799.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P327467 | AK144918.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114526 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139478 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09495187 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09658894 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09658890 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09658903 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09658885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767873 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767867 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767875 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767872 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767868 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767869 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767871 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767870 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27767874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27767902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767906 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09550362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767834 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767904 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09645261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767896 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09722646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08992435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767918 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09658900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767902 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09300884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767837 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767832 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09722638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09446709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767830 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767831 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09658885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27767882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09372778 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375440 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375441 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10375442 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417564_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10375439 | - | - |
| Mouse Genome 430 2.0 Array | 1417564_at | - | - |
| Mouse Genome 430 2.0 Array | 1417564_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66213_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66213_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100003627.mm.1 | NM_001104530.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003627.mm.1 | NM_001104530.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003627.mm.1 | NM_025426.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003627.mm.1 | NM_025426.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100003627.mm.1 | TC1100000473.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417564_PM_at | - | - |
| Mouse Expression Set 430 | 1417564_at | - | - |
| Murine Genome U74v2 Set | 168695_i_at | - | - |
| Murine Genome U74v2 Set | 168695_PM_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417564_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417564_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417564_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417564_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 452858 | - | - |
| ABI Mouse Genome Survey Microarray | 930034 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4850092 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4850092 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850092 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225666 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225666 | - | - |
| mouse-6 v1.1 (Illumina) | scl41644.4_98-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850092 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl41644.4_98-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850092 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2832091 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225666 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225666 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2832091 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl41644.4_98-S | - | - |

Med7
Genomic Map