Assay Details
| Gene Symbol: Mrpl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025606.3 | NP_079882.2 | 2 - 3 | 171 | 109 | |
| GenBank mRNA | AB049643.1 | - | 2 - 3 | 171 | 109 | |
| AK004034.1 | - | 2 - 3 | 162 | 109 | ||
| AK009687.1 | - | 2 - 3 | 157 | 109 | ||
| AK011971.1 | - | 2 - 3 | 178 | 109 | ||
| AK165616.1 | - | 2 - 3 | 204 | 109 | ||
| BC044721.1 | - | 2 - 3 | 195 | 6476049 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl16
Genomic Map