Assay Details
| Gene Symbol: Pgd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081274.1 | NP_001074743.1 | 8 - 9 | 960 | 70 | |
| GenBank mRNA | AK002894.1 | - | 8 - 9 | 922 | 70 | |
| AK004217.1 | - | 1 - 2 | 164 | 70 | ||
| AK145602.1 | - | 8 - 9 | 924 | 70 | ||
| AK150210.1 | - | 8 - 9 | 923 | 70 | ||
| AK150490.1 | - | 8 - 9 | 919 | 70 | ||
| AK151380.1 | - | 8 - 9 | 919 | 70 | ||
| AK151993.1 | - | 8 - 9 | 919 | 70 | ||
| AK152758.1 | - | 8 - 9 | 919 | 70 | ||
| AK152894.1 | - | 8 - 9 | 919 | 70 | ||
| AK153409.1 | - | 8 - 9 | 916 | 70 | ||
| AK155027.1 | - | 8 - 9 | 919 | 70 | ||
| AK166733.1 | - | 8 - 9 | 914 | 70 | ||
| AK166947.1 | - | 8 - 9 | 896 | 70 | ||
| AK167215.1 | - | 8 - 9 | 914 | 70 | ||
| AK168251.1 | - | 8 - 9 | 919 | 70 | ||
| AK180298.1 | - | 1 - 2 | 121 | 70 | ||
| BC008646.1 | - | 8 - 9 | 891 | 3155147 | 70 | |
| BC011329.1 | - | 8 - 9 | 909 | 3984654 | 70 | |
| BC014793.1 | - | 8 - 9 | 928 | 3155100 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pgd
Genomic Map