Assay Details
| Gene Symbol: Pnpla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163689.1 | NP_001157161.1 | 2 - 3 | 521 | 70 | |
| NM_025802.3 | NP_080078.2 | 2 - 3 | 521 | 70 | ||
| NR_028142.1 | 3 - 4 | 603 | 70 | |||
| GenBank mRNA | AK002826.1 | - | 2 - 3 | 503 | 70 | |
| AK031609.1 | - | 2 - 3 | 479 | 70 | ||
| AK076464.1 | - | 2 - 3 | 305 | 70 | ||
| AK085926.1 | - | 2 - 3 | 505 | 70 | ||
| AK150184.1 | - | 2 - 3 | 503 | 70 | ||
| AY510273.1 | - | 2 - 3 | 502 | 70 | ||
| AY731699.1 | - | 2 - 3 | 502 | 70 | ||
| AY894805.1 | - | 2 - 3 | 422 | 70 | ||
| BC019188.1 | - | 3 - 4 | 585 | 5027544 | 70 | |
| BC044737.1 | - | 3 - 4 | 567 | 70 | ||
| BC044781.2 | - | 3 - 4 | 567 | 5151911 | 70 | |
| BC054825.1 | - | 3 - 4 | 562 | 70 | ||
| BC064747.1 | - | 2 - 3 | 493 | 30024535 | 70 | |
| CF725975.1 | - | 2 - 3 | 511 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnpla2
Genomic Map