Assay Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025813.3 | NP_080089.1 | 9 - 10 | 910 | 71 | |
| GenBank mRNA | AK004586.1 | - | 9 - 10 | 911 | 71 | |
| AK032688.1 | - | 9 - 10 | 878 | 71 | ||
| AK075859.1 | - | 9 - 10 | 940 | 71 | ||
| AK135235.1 | - | 9 - 10 | 911 | 71 | ||
| AK136108.1 | - | 9 - 10 | 902 | 71 | ||
| AK152444.1 | - | 9 - 10 | 932 | 71 | ||
| AK159554.1 | - | 9 - 10 | 938 | 71 | ||
| BC024891.1 | - | 9 - 10 | 892 | 3992247 | 71 | |
More Information
Set Membership: |
|

Mfsd1
Genomic Map