Assay Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025813.3 | NP_080089.1 | 11 - 12 | 1125 | 70 | |
| GenBank mRNA | AK004586.1 | - | 11 - 12 | 1126 | 70 | |
| AK032688.1 | - | 11 - 12 | 1093 | 70 | ||
| AK075859.1 | - | 11 - 12 | 1155 | 70 | ||
| AK135235.1 | - | 11 - 12 | 1126 | 70 | ||
| AK136108.1 | - | 11 - 12 | 1117 | 70 | ||
| AK152444.1 | - | 11 - 12 | 1147 | 70 | ||
| AK159554.1 | - | 11 - 12 | 1153 | 70 | ||
| AK166645.1 | - | 5 - 6 | 517 | 70 | ||
| AK191801.1 | - | 1 - 2 | 57 | 70 | ||
| BC024891.1 | - | 11 - 12 | 1107 | 3992247 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd1
Genomic Map