Assay Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025813.3 | NP_080089.1 | 13 - 14 | 1336 | 67 | |
| GenBank mRNA | AK004586.1 | - | 13 - 14 | 1337 | 67 | |
| AK032688.1 | - | 13 - 14 | 1304 | 67 | ||
| AK075859.1 | - | 13 - 14 | 1366 | 67 | ||
| AK135235.1 | - | 13 - 14 | 1337 | 67 | ||
| AK136108.1 | - | 13 - 14 | 1328 | 67 | ||
| AK152444.1 | - | 13 - 14 | 1358 | 67 | ||
| AK159554.1 | - | 13 - 14 | 1364 | 67 | ||
| AK166645.1 | - | 7 - 8 | 728 | 67 | ||
| AK191801.1 | - | 3 - 4 | 268 | 67 | ||
| AK214765.1 | - | 2 - 3 | 134 | 67 | ||
| BC024891.1 | - | 13 - 14 | 1318 | 3992247 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd1
Genomic Map