Assay Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025813.3 | NP_080089.1 | 14 - 15 | 1387 | 71 | |
| GenBank mRNA | AK004586.1 | - | 14 - 15 | 1388 | 71 | |
| AK032688.1 | - | 14 - 15 | 1355 | 71 | ||
| AK075859.1 | - | 14 - 15 | 1417 | 71 | ||
| AK135235.1 | - | 14 - 15 | 1388 | 71 | ||
| AK136108.1 | - | 14 - 15 | 1379 | 71 | ||
| AK152444.1 | - | 14 - 15 | 1409 | 71 | ||
| AK159554.1 | - | 14 - 15 | 1415 | 71 | ||
| AK166645.1 | - | 8 - 9 | 779 | 71 | ||
| AK191801.1 | - | 4 - 5 | 319 | 71 | ||
| AK214765.1 | - | 3 - 4 | 185 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd1
Genomic Map