Assay Details
| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026168.4 | NP_080444.1 | 3 - 4 | 660 | 84 | |
| NM_026355.3 | NP_080631.1 | 3 - 4 | 660 | 84 | ||
| GenBank mRNA | AK004661.1 | - | 3 - 4 | 324 | 84 | |
| AK007498.1 | - | 3 - 4 | 309 | 84 | ||
| AK008971.1 | - | 3 - 4 | 316 | 84 | ||
| AK009483.1 | - | 3 - 4 | 312 | 84 | ||
| AK010529.1 | - | 3 - 4 | 309 | 84 | ||
| AK014413.1 | - | 3 - 4 | 316 | 84 | ||
| AK016275.1 | - | 3 - 4 | 661 | 84 | ||
| AK017876.1 | - | 3 - 4 | 344 | 84 | ||
| AK030045.1 | - | 3 - 4 | 310 | 84 | ||
| AK086727.1 | - | 3 - 4 | 312 | 84 | ||
| AK167391.1 | - | 3 - 4 | 313 | 84 | ||
| AK214649.1 | - | 2 - 3 | 133 | 84 | ||
| BC006895.1 | - | 3 - 4 | 278 | 2651322 | 84 | |
| BC018188.1 | - | 3 - 4 | 312 | 3711066 | 84 | |
| BC026558.1 | - | 3 - 4 | 272 | 5030661 | 84 | |
| BC064749.1 | - | 3 - 4 | 373 | 30046220 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ergic2
Genomic Map