Assay Details
| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286560.1 | NP_001273489.1 | 3 - 4 | 595 | 86 | |
| NM_026168.4 | NP_080444.1 | 4 - 5 | 704 | 86 | ||
| NM_026355.3 | NP_080631.1 | 4 - 5 | 704 | 86 | ||
| GenBank mRNA | AK004661.1 | - | 4 - 5 | 368 | 86 | |
| AK007498.1 | - | 4 - 5 | 353 | 86 | ||
| AK008971.1 | - | 4 - 5 | 360 | 86 | ||
| AK009483.1 | - | 4 - 5 | 356 | 86 | ||
| AK010529.1 | - | 4 - 5 | 353 | 86 | ||
| AK014413.1 | - | 4 - 5 | 360 | 86 | ||
| AK016275.1 | - | 4 - 5 | 705 | 86 | ||
| AK017876.1 | - | 4 - 5 | 388 | 86 | ||
| AK030045.1 | - | 4 - 5 | 354 | 86 | ||
| AK086727.1 | - | 4 - 5 | 356 | 86 | ||
| AK162267.1 | - | 3 - 4 | 328 | 86 | ||
| AK167391.1 | - | 4 - 5 | 357 | 86 | ||
| AK214649.1 | - | 3 - 4 | 177 | 86 | ||
| BC006895.1 | - | 4 - 5 | 322 | 2651322 | 86 | |
| BC018188.1 | - | 4 - 5 | 356 | 3711066 | 86 | |
| BC026558.1 | - | 4 - 5 | 316 | 5030661 | 86 | |
| BC064749.1 | - | 4 - 5 | 417 | 30046220 | 86 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286560.1 transcripts.
Set Membership: |
|

Ergic2
Genomic Map