Assay Details
| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286560.1 | NP_001273489.1 | 4 - 5 | 669 | 82 | |
| NM_026168.4 | NP_080444.1 | 5 - 6 | 778 | 82 | ||
| NM_026355.3 | NP_080631.1 | 5 - 6 | 778 | 82 | ||
| GenBank mRNA | AK004661.1 | - | 5 - 6 | 442 | 82 | |
| AK007498.1 | - | 5 - 6 | 427 | 82 | ||
| AK008971.1 | - | 5 - 6 | 434 | 82 | ||
| AK009483.1 | - | 5 - 6 | 430 | 82 | ||
| AK010529.1 | - | 5 - 6 | 427 | 82 | ||
| AK016275.1 | - | 5 - 6 | 779 | 82 | ||
| AK017876.1 | - | 5 - 6 | 462 | 82 | ||
| AK030045.1 | - | 5 - 6 | 428 | 82 | ||
| AK086727.1 | - | 5 - 6 | 430 | 82 | ||
| AK135968.1 | - | 2 - 3 | 115 | 82 | ||
| AK162267.1 | - | 4 - 5 | 402 | 82 | ||
| AK167391.1 | - | 5 - 6 | 431 | 82 | ||
| BC006895.1 | - | 5 - 6 | 396 | 2651322 | 82 | |
| BC018188.1 | - | 5 - 6 | 430 | 3711066 | 82 | |
| BC026558.1 | - | 5 - 6 | 390 | 5030661 | 82 | |
| BC064749.1 | - | 5 - 6 | 491 | 30046220 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ergic2
Genomic Map