Assay Details
| Gene Symbol: Ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286560.1 | NP_001273489.1 | 5 - 6 | 704 | 62 | |
| NM_026168.4 | NP_080444.1 | 6 - 7 | 813 | 62 | ||
| NM_026355.3 | NP_080631.1 | 6 - 7 | 813 | 62 | ||
| GenBank mRNA | AK004661.1 | - | 6 - 7 | 477 | 62 | |
| AK007498.1 | - | 6 - 7 | 462 | 62 | ||
| AK008971.1 | - | 6 - 7 | 469 | 62 | ||
| AK009483.1 | - | 6 - 7 | 465 | 62 | ||
| AK010529.1 | - | 6 - 7 | 462 | 62 | ||
| AK016275.1 | - | 6 - 7 | 814 | 62 | ||
| AK017876.1 | - | 6 - 7 | 497 | 62 | ||
| AK030045.1 | - | 6 - 7 | 463 | 62 | ||
| AK086727.1 | - | 6 - 7 | 465 | 62 | ||
| AK135968.1 | - | 3 - 4 | 150 | 62 | ||
| AK162267.1 | - | 5 - 6 | 437 | 62 | ||
| AK167391.1 | - | 6 - 7 | 466 | 62 | ||
| BC006895.1 | - | 6 - 7 | 431 | 2651322 | 62 | |
| BC018188.1 | - | 6 - 7 | 465 | 3711066 | 62 | |
| BC026558.1 | - | 6 - 7 | 425 | 5030661 | 62 | |
| BC064749.1 | - | 6 - 7 | 526 | 30046220 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ergic2
Genomic Map