Assay Details
| Gene Symbol: Frmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026169.4 | NP_080445.1 | 1 - 2 | 119 | 71 | |
| GenBank mRNA | AK004603.2 | - | 1 - 2 | 142 | 71 | |
| AK049224.1 | - | 1 - 2 | 134 | 71 | ||
| AK049233.1 | - | 1 - 2 | 134 | 71 | ||
| AK083902.1 | - | 1 - 2 | 121 | 71 | ||
| AK088639.1 | - | 1 - 2 | 131 | 71 | ||
| AK148434.1 | - | 131 | 71 | |||
| AK168669.1 | - | 1 - 2 | 131 | 71 | ||
| AK206183.1 | - | 1 - 2 | 83 | 71 | ||
| AK214415.1 | - | 1 - 2 | 83 | 71 | ||
| BC004765.1 | - | 1 - 2 | 187 | 3156148 | 71 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK004603.2,AK049224.1,AK049233.1,AK083902.1,AK088639.1,AK148434.1,BC004765.1,NM_026169.4 transcripts.
Set Membership: |
|

Frmd8
Genomic Map