Assay Details
| Gene Symbol: Frmd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026169.4 | NP_080445.1 | 6 - 7 | 706 | 80 | |
| GenBank mRNA | AK004603.2 | - | 6 - 7 | 729 | 80 | |
| AK049224.1 | - | 6 - 7 | 721 | 80 | ||
| AK049233.1 | - | 6 - 7 | 721 | 80 | ||
| AK083902.1 | - | 6 - 7 | 708 | 80 | ||
| AK088639.1 | - | 6 - 7 | 718 | 80 | ||
| AK148434.1 | - | 718 | 80 | |||
| AK168669.1 | - | 6 - 7 | 717 | 80 | ||
| BC004765.1 | - | 6 - 7 | 774 | 3156148 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Frmd8
Genomic Map