Assay Details
| Gene Symbol: Get4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163316.1 | NP_001156788.1 | 6 - 7 | 703 | 73 | |
| NM_026269.2 | NP_080545.2 | 6 - 7 | 894 | 73 | ||
| GenBank mRNA | AK003551.1 | - | 6 - 7 | 843 | 73 | |
| AK185239.1 | - | 1 - 2 | 66 | 73 | ||
| AK212006.1 | - | 1 - 2 | 65 | 73 | ||
| BC019557.1 | - | 6 - 7 | 685 | 4163641 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Get4
Genomic Map