Assay Details
| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026470.3 | NP_080746.3 | 2 - 3 | 390 | 62 | |
| GenBank mRNA | AK051574.1 | - | 2 - 3 | 391 | 62 | |
| AK053170.1 | - | 2 - 3 | 364 | 62 | ||
| AK132771.1 | - | 2 - 3 | 364 | 62 | ||
| AK153098.1 | - | 2 - 3 | 267 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spata6
Genomic Map