Assay Details
| Gene Symbol: Spata6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026470.3 | NP_080746.3 | 9 - 10 | 1099 | 73 | |
| GenBank mRNA | AF032967.1 | - | 3 - 4 | 406 | 74 | |
| AK005819.1 | - | 4 - 5 | 486 | 73 | ||
| AK006861.1 | - | 4 - 5 | 471 | 73 | ||
| AK051574.1 | - | 9 - 10 | 1100 | 73 | ||
| AK132771.1 | - | 9 - 10 | 1073 | 73 | ||
| AK153098.1 | - | 9 - 10 | 976 | 73 | ||
| AK183508.1 | - | 3 - 4 | 180 | 73 | ||
| BC028429.1 | - | 4 - 5 | 499 | 73 | ||
| BC040024.1 | - | 4 - 5 | 499 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spata6
Genomic Map