Assay Details
| Gene Symbol: Golt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026680.4 | NP_080956.1 | 3 - 4 | 363 | 65 | |
| GenBank mRNA | AK002581.1 | - | 3 - 4 | 371 | 65 | |
| AK141378.1 | - | 3 - 4 | 364 | 65 | ||
| AY707855.1 | - | 3 - 4 | 386 | 65 | ||
| BC024448.1 | - | 3 - 4 | 386 | 4974299 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Golt1a
Genomic Map