| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10349910 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10349915 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10349908 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10349906 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10349905 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10349909 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10349911 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10349914 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10349907 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10349912 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1425159_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10349904 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10349913 |
- |
- |
| Mouse Genome 430 2.0 Array |
1425159_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1425159_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
68338_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
68338_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0100009867.mm.1 |
NM_026680.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009867.mm.1 |
NM_026680.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009867.mm.1 |
TC0100001205.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009868.mm.1 |
NM_026680.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009868.mm.1 |
NM_026680.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009868.mm.1 |
TC0100001205.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009869.mm.1 |
NM_026680.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009869.mm.1 |
NM_026680.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009869.mm.1 |
TC0100001205.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100009866.mm.1 |
NM_026680.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009866.mm.1 |
NM_026680.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100009866.mm.1 |
TC0100001205.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1425159_PM_at |
- |
- |
| Mouse Expression Set 430 |
1425159_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1425159_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1425159_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1425159_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1425159_at |
- |
- |